Methyl-3-(trifluoromethyl)phenylacetate 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl 2-(3-(trifluoromethyl)phenyl)acetate
H32883.03
H32883.06
H32883.14
H32883.03EA
38.2
EUR
H32883.03
H32883.06
H32883.14
Methyl-3-(trifluoromethyl)phenylacetate 95%
Methyl-3-(trifluoromethyl)phenylacetate
Purity: 95%
CAS No.: 62451-84-7
Molecular Formula: C10H9F3O2
Formula Weight: 218.18
Refractive Index: 1.4500
Boiling Point: 211°
MDL No.: MFCD04039762
Formel:
C₁₀H₉F₃O₂ Siedepunkt: 211 °C Dichte: 1.24 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04039762 CAS-Nummer: 62451-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...