2,2,2-Trimethylthioacetamide 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H32092.03
H32092.09
H32092.03EA
53.6
EUR
H32092.03
H32092.09
2,2,2-Trimethylthioacetamide 97%
2,2,2-Trimethylthioacetamide
Purity: 97%
CAS No.: 630-22-8
Molecular Formula: C5H11NS
Formula Weight: 117.21
Melting Point: 113-117°
MDL No.: MFCD09742834
Formel:
C₅H₁₁NS Schmelzpunkt: 113…117 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09742834 CAS-Nummer: 630-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...