4-n-Dodecylbenzoic acid 98%

Lieferant: Thermo Fisher Scientific

H31983.03 H31983.06
H31983.03EA 62.2 EUR
H31983.03 H31983.06
4-n-Dodecylbenzoic acid 98%
4-n-Dodecylbenzoic acid

Purity: 98% CAS No.: 21021-55-6 Molecular Formula: C19H30O2 Formula Weight: 290.45 MDL No.: MFCD04972235

Formel: C₁₉H₃₀O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04972235
CAS-Nummer: 21021-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR