3-(1H-Tetrazol-5-yl)benzoic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(1H-Tetrazol-5-yl)benzoic Acid
H27581.MD
H27581.03
H27581.MDEA
33.5
EUR
H27581.MD
H27581.03
3-(1H-Tetrazol-5-yl)benzoic acid 97%
3-(1H-Tetrazol-5-yl)benzoic acid
Purity: 97%
CAS No.: 73096-39-6
Molecular Formula: C8H6N4O2
Formula Weight: 190.16
Melting Point: 284-286°
MDL No.: MFCD03821131
Formel:
C₈H₆N₄O₂ Schmelzpunkt: 284…286 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03821131 CAS-Nummer: 73096-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...