1-Methyl-4-piperidin-4-ylpiperazine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Methyl-4-(piperidin-4-yl)piperazine

H27408.06 H27408.03
H27408.06EA 166 EUR
H27408.06 H27408.03
1-Methyl-4-piperidin-4-ylpiperazine 98%
1-Methyl-4-piperidin-4-ylpiperazine

Purity: 98% CAS No.: 53617-36-0 Molecular Formula: C10H21N3 Formula Weight: 183.30 Melting Point: 53-56° UN No.: UN3259 MDL No.: MFCD03274729

Formel: C₁₀H₂₁N₃
Schmelzpunkt: 53…56 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03274729
CAS-Nummer: 53617-36-0
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR