L-m-Tyrosine ≥97%
Lieferant: Thermo Fisher Scientific
H27224.MD
H27224.03
H27224.MDEA
111
EUR
H27224.MD
H27224.03
L-m-Tyrosine ≥97%
L-m-Tyrosine
Purity: 97+%
CAS No.: 587-33-7
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: 260-270°
MDL No.: MFCD00063059
Formel:
C₉H₁₁NO₃ Schmelzpunkt: 260…270 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00063059 CAS-Nummer: 587-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...