(S)-2-Methylpropane-2-sulphinamide 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-tert-Butylsulphinamide, (S)-(-)-tert-Butylsulphinamide, (S)-(-)-t-Butylsulphinamide, (S)-(-)-2-Methyl-2-propanesulphinamide, (S)-(-)-tert-Butyl sulphinamide
H27115.14
H27115.06
H27115.03
H27115.14EA
936
EUR
H27115.14
H27115.06
H27115.03
(S)-2-Methylpropane-2-sulphinamide 97%
(S)-2-Methylpropane-2-sulphinamide
Purity: 97%
CAS No.: 343338-28-3
Molecular Formula: C4H11NOS
Formula Weight: 121.20
Storage Temperature: 2°C to 8°C
Melting Point: 97-101°
MDL No.: MFCD05861480
Formel:
C₄H₁₁NOS Schmelzpunkt: 97…101 °C |
MDL Number:
MFCD05861480 CAS-Nummer: 343338-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...