Di-(N-succinimidyl)-carbonat ≥85% remainder N-Hydroxysuccinimide, tech.

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate

H26629.03 H26629.06 H26629.14
H26629.03EA 27.8 EUR
H26629.03 H26629.06 H26629.14
Di-(N-succinimidyl)-carbonat ≥85% remainder N-Hydroxysuccinimide, tech.
Di(N-succinimidyl) carbonate

Purity: 85% CAS No.: 74124-79-1 Molecular Formula: C9H8N2O7 Formula Weight: 256.17 Melting Point: 190° dec. Beilstein Reference: 1499137 MDL No.: MFCD00009767

Formel: C₉H₈N₂O₇
Schmelzpunkt: 190 °C (dec.)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00009767
CAS-Nummer: 74124-79-1
EINECS: 277-730-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR