Bis(toluene-4-sulphonyl)methane 97%
Lieferant: Thermo Fisher Scientific
H26617.03
H26617.06
H26617.03EA
95.8
EUR
H26617.03
H26617.06
Bis(toluene-4-sulphonyl)methane 97%
Bis(toluene-4-sulphonyl)methane
Purity: 97%
CAS No.: 15310-28-8
Molecular Formula: C15H16O4S2
Formula Weight: 324.42
Melting Point: 137-139°
MDL No.: MFCD00032255
Formel:
C₁₅H₁₆O₄S₂ Schmelzpunkt: 137…139 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00032255 CAS-Nummer: 15310-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...