2,6-Pyridinedicarbonyl dichloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,6-Pyridinedicarboxylicacidchloride, 2,6-Pyridinedicarboxylic acid chloride

H26570.03 H26570.06 H26570.14
H26570.03EA 24.2 EUR
H26570.03 H26570.06 H26570.14
2,6-Pyridinedicarbonyl dichloride 97%
2,6-Pyridinedicarbonyl dichloride

Purity: 98% CAS No.: 3739-94-4 Molecular Formula: C7H3Cl2NO2 Formula Weight: 204.01 Odour: Irritating Melting Point: 57-61° Boiling Point: 284° Beilstein Reference: 131556 UN No.: UN3261 MDL No.: MFCD00006289

Formel: C₇H₃Cl₂NO₂
Siedepunkt: 284 °C
Schmelzpunkt: 57…61 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006289
CAS-Nummer: 3739-94-4
EINECS: 223-125-4
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR