5-Carbethoxyuracil 98%

Lieferant: Thermo Fisher Scientific

H26507.03 H26507.06
H26507.03EA 25.5 EUR
H26507.03 H26507.06
5-Carbethoxyuracil 98%
5-Carbethoxyuracil

Purity: 98% CAS No.: 28485-17-8 Molecular Formula: C7H8N2O4 Formula Weight: 184.15 MDL No.: MFCD00057337

Formel: C₇H₈N₂O₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00057337
CAS-Nummer: 28485-17-8
EINECS: 249-053-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR