N-Boc-L-prolinal 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-1-Boc-2-Formylpyrrolidine

H26495.06 H26495.03
H26495.06EA 480 EUR
H26495.06 H26495.03
N-Boc-L-prolinal 96%
N-Boc-L-prolinal

Purity: 96% CAS No.: 69610-41-9 Molecular Formula: C10H17NO3 Formula Weight: 199.25 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4589 Boiling Point: 228° Optical Rotation: -95° (c=1 in chloroform) MDL No.: MFCD00274186

Formel: C₁₀H₁₇NO₃
Siedepunkt: 228 °C
Dichte: 1.06
Flammpunkt: >110 °C (230 °F)
MDL Number: MFCD00274186
CAS-Nummer: 69610-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR