Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester
Purity: 97%
CAS No.: 571-55-1
Molecular Formula: C9H11F3O4
Formula Weight: 240.18
Flash Point: 104°(220°F)
Refractive Index: 1.4290
Boiling Point: 80-82°/1mm
MDL No.: MFCD02677683
Formel:
C₉H₁₁F₃O₄ Siedepunkt: 80…82 °C (1 mmHg) Dichte: 1.235 Flammpunkt: 104 °C (220 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02677683 CAS-Nummer: 571-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...