Albendazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
[5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester, Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate
H25925.09
H25925.18
H25925.22
H25925.09EA
96.7
EUR
H25925.09
H25925.18
H25925.22
Albendazol ≥98%
Albendazol
Purity: 98+%
CAS No.: 54965-21-8
Molecular Formula: C12H15N3O2S
Formula Weight: 265.34
Melting Point: 208-210°
MDL No.: MFCD00083232
Formel:
C₁₂H₁₅N₃O₂S Schmelzpunkt: 208…210 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00083232 CAS-Nummer: 54965-21-8 EINECS: 259-414-7 Merck Index: 14,210 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...