Amberlite® IRA-402 (Cl) Ionenaustauscher
Lieferant: Thermo Fisher Scientific
Synonyme:
Amberlite ® IRA402 chlorine type
B25556.36
B25556.22
B25556.0E
B25556.36EA
70.6
EUR
B25556.36
B25556.22
B25556.0E
Amberlite® IRA-402 (Cl) Ionenaustauscher
Amberlite® IRA-402 (Cl)
CAS No.: 52439-77-7
MDL No.: MFCD01864002
Dichte:
1.07 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00145564 CAS-Nummer: 52439-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...