4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(p-Tolyl)-2,2:6,2-terpyridine
B25509.02
B25509.77
B25509.02EA
156
EUR
B25509.02
B25509.77
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine
Purity: 98%
CAS No.: 89972-77-0
Molecular Formula: C22H17N3
Formula Weight: 323.40
Melting Point: 167-170°
MDL No.: MFCD06796977
Formel:
C₂₂H₁₇N₃ Molekulargewicht: 323.40 g/mol Schmelzpunkt: 167…170 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06796977 CAS-Nummer: 89972-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...