O,O'-Di-2-pyridyl thiocarbonate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Di-2-Pyridyl Thionocarbonate, Thiocarbonic Acid O,O'-Di-2-pyridyl Ester
B22451.03
B22451.06
B22451.14
B22451.03EA
75.4
EUR
B22451.03
B22451.06
B22451.14
O,O'-Di-2-pyridyl thiocarbonate 98%
O,O'-Di-2-pyridyl thiocarbonate
Purity: 98%
CAS No.: 96989-50-3
Molecular Formula: C11H8N2O2S
Formula Weight: 232.26
Storage Temperature: -30°C to -10°C
Melting Point: 96-99°
MDL No.: MFCD00074870
Formel:
C₁₁H₈N₂O₂S Molekulargewicht: 232.26 g/mol Schmelzpunkt: 96…99 °C |
MDL Number:
MFCD00074870 CAS-Nummer: 96989-50-3 |