Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}tyrosinat 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester
B22164.06
B22164.14
B22164.22
B22164.06EA
70.9
EUR
B22164.06
B22164.14
B22164.22
Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}tyrosinat 99%
Boc-L-tyrosine methyl ester
Purity: 99%
CAS No.: 4326-36-7
Molecular Formula: C15H21NO5
Formula Weight: 295.34
Melting Point: 105-108°
Optical Rotation: +50° (c=1 chloroform)
MDL No.: MFCD00191181
Formel:
C₁₅H₂₁NO₅ Molekulargewicht: 295.3 g/mol Schmelzpunkt: 105…108 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191181 CAS-Nummer: 4326-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...