Aminoacetonitril-Sulfat 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Aminoacetonitrile hemisulphate, Glycinonitrile sulphate, Cyanomethylamine sulphate
B22155.14
B22155.22
B22155.36
B22155.14EA
34.1
EUR
B22155.14
B22155.22
B22155.36
Aminoacetonitril-Sulfat 97%
Aminoacetonitrile sulphate
Purity: 97%
CAS No.: 5466-22-8
Molecular Formula: C4H8N8.H2SO4
Formula Weight: 210.21
Odour: Odourless
Melting Point: ca 160° dec.
Beilstein Reference: 3916094
UN No.: UN3439
MDL No.: MFCD00012851
Formel:
C₄H₁₀N₄O₄S Schmelzpunkt: ∼ 160 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012851 CAS-Nummer: 5466-22-8 EINECS: 226-772-0 ADR: 6.1,III Merck Index: 14,412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...