Oxalessigsäure-diethylester Natriumsalz 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium diethyl oxalacetate, Sodium Diethyl Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt

B21486.22 B21486.36
B21486.22EA 45.9 EUR
B21486.22 B21486.36
Oxalessigsäure-diethylester Natriumsalz 95%
Diethyl oxalacetate sodium salt

Purity: 95% CAS No.: 40876-98-0 Molecular Formula: C8H11NaO5 Formula Weight: 210.16 Melting Point: ca 185° Beilstein Reference: 4279609 MDL No.: MFCD00035571

Formel: C₈H₁₂O₅
Schmelzpunkt: ∼ 185 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00035571
CAS-Nummer: 40876-98-0
EINECS: 255-122-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR