Guanylureasulfat 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Dicyandiamidine Sulphate

A19106.30 A19106.09 A19106.18
A19106.30EA 407 EUR
A19106.30 A19106.09 A19106.18
Guanylureasulfat 97%
Guanylurea sulphate

Purity: 97% CAS No.: 591-01-5 Molecular Formula: C4H12N8O2.H2SO4 Formula Weight: 302.27 Melting Point: ca 195° dec. Beilstein Reference: 6099371 MDL No.: MFCD00035565

Formel: C₂H₈N₄O₅S
Molekulargewicht: 200.17 g/mol
Schmelzpunkt: ∼ 195 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00035565
CAS-Nummer: 591-01-5
EINECS: 209-697-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR