Salbutamol sulphate 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bronchospray, Proventil-HFA, Salbutamol hemisulphate, Albuterol hemissulphate, 2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulphate, Albutarol sulphate, 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulphate

A18544.03 A18544.06
A18544.03EA 43.1 EUR
A18544.03 A18544.06
Salbutamol sulphate 99%
Salbutamol sulphate

Purity: 99% CAS No.: 51022-70-9 Molecular Formula: C26H42N2O6.H2SO4 Formula Weight: 576.70 MDL No.: MFCD00055200

Formel: (C₁₃H₂₁NO₃)₂·H₂SO₄
Molekulargewicht: 576.702 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00055200
CAS-Nummer: 51022-70-9
EINECS: 256-916-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR