N-Acetyl-ε-caprolactam 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
N-Acetyl-2-oxohexamethyleneimine, N-Acetylcaprolactam
A18257.14
A18257.22
A18257.14EA
40.1
EUR
A18257.14
A18257.22
N-Acetyl-ε-caprolactam 99%
N-Acetyl-epsilon-caprolactam
Purity: 99%
CAS No.: 1888-91-1
Molecular Formula: C8H13NO2
Formula Weight: 155.20
Flash Point: 122°(252°F)
Refractive Index: 1.4890
Boiling Point: 124°/12mm
Beilstein Reference: 119598
MDL No.: MFCD00003262
Formel:
C₈H₁₃NO₂ Siedepunkt: 124 °C (12 mmHg) Dichte: 1.094 Flammpunkt: 122 °C (252 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003262 CAS-Nummer: 1888-91-1 EINECS: 217-565-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...