Copper(II) phosphate 98%

Lieferant: Thermo Fisher Scientific
Achtung

A17360.0E A17360.22 A17360.36
A17360.0EEA 1040 EUR
A17360.0E A17360.22 A17360.36
Copper(II) phosphate 98%
Copper(II) phosphate
Formel: Cu₃O₈P₂
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00036268
CAS-Nummer: 7798-23-4
EINECS: 232-254-5
Merck Index: 14,2648

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR