Diphenylphosphinamide 98%

Lieferant: Thermo Fisher Scientific
Achtung

A16657.14 A16657.06
A16657.14EA 504 EUR
A16657.14 A16657.06
Diphenylphosphinamide 98%
Diphenylphosphinamide

Purity: 98% CAS No.: 5994-87-6 Molecular Formula: C12H12NOP Formula Weight: 217.21 Melting Point: 160-162° MDL No.: MFCD00014766

Formel: C₁₂H₁₂NOP
Schmelzpunkt: 160…162 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014766
CAS-Nummer: 5994-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR