3-Hydroxy-2-naphthohydrazid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-2-naphthohydrazide, 2-Hydroxy-3-naphthoic Acid Hydrazide
A16192.14
A16192.22
A16192.14EA
75.5
EUR
A16192.14
A16192.22
3-Hydroxy-2-naphthohydrazid 98%
3-Hydroxy-2-naphthohydrazid
Purity: 98%
CAS No.: 5341-58-2
Molecular Formula: C11H10N2O2
Formula Weight: 202.21
Melting Point: ca 205° dec.
Beilstein Reference: 395153
MDL No.: MFCD00004097
Formel:
C₁₁H₁₀N₂O₂ Schmelzpunkt: ∼ 205 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004097 CAS-Nummer: 5341-58-2 EINECS: 226-282-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...