5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
5,6-Dimethyl-2-benzimidazolinone
A15421.03
A15421.06
A15421.03EA
49.7
EUR
A15421.03
A15421.06
5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one 98%
5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Purity: 98%
CAS No.: 2033-30-9
Molecular Formula: C9H10N2O
Formula Weight: 162.19
Melting Point:
Formel:
C₉H₁₀N₂O Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051725 CAS-Nummer: 2033-30-9 EINECS: 217-993-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...