DL-2,4-Diaminobutyric acid dihydrochloride 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-DL-Dab·2HCl, H-DL-Dab.2HCl
A15408.03
A15408.06
A15408.14
A15408.03EA
62.7
EUR
A15408.03
A15408.06
A15408.14
DL-2,4-Diaminobutyric acid dihydrochloride 99%
DL-2,4-Diaminobutyric acid dihydrochloride
Purity: 99%
CAS No.: 65427-54-5
Molecular Formula: C4H10N2O2.2HCl
Formula Weight: 191.06
Melting Point: ca 198° dec.
Beilstein Reference: 4708499
MDL No.: MFCD00012910
Formel:
C₄H₁₀N₂O₂·₂HCl Schmelzpunkt: ∼ 198 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012910 CAS-Nummer: 65427-54-5 EINECS: 265-774-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...