Diethyl phenylthiomethylphosphonate 96%
Lieferant: Thermo Fisher Scientific
A13660.09
A13660.18
A13660.09EA
95.5
EUR
A13660.09
A13660.18
Diethyl phenylthiomethylphosphonate 96%
Diethyl phenylthiomethylphosphonate
Purity: 96%
CAS No.: 38066-16-9
Molecular Formula: C11H17O3PS
Formula Weight: 260.29
Refractive Index: 1.5340
Boiling Point: 146-148°/0.5mm
Beilstein Reference: 1246215
MDL No.: MFCD00015130
Formel:
C₁₁H₁₇O₃PS Siedepunkt: 146…148 °C (0.5 mmHg) Dichte: 1.174 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015130 CAS-Nummer: 38066-16-9 EINECS: 253-770-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...