DL-Alanine methyl ester hydrochloride ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme: H-DL-Ala-OMe.HCl, H-DL-Ala-OMe·HCl
A12599.06 A12599.14
A12599.06EA 35.4 EUR
A12599.06 A12599.14
DL-Alanine methyl ester hydrochloride ≥98%
DL-Alanine methyl ester hydrochloride
Purity: 98+% CAS No.: 13515-97-4 Molecular Formula: C4H9NO2.HCl Formula Weight: 139.58 Storage Temperature: 2°C to 8°C Melting Point: ca 155° dec. Beilstein Reference: 3619264 MDL No.: MFCD00035523
Formel: C₄H₉NO₂
Molekulargewicht: 139.58 g/mol
Schmelzpunkt: ∼ 155 °C (Zers.)
MDL Number: MFCD00035523
CAS-Nummer: 13515-97-4
EINECS: 236-854-8