N-Benzoyl-N-phenylhydroxylamine 98%
Lieferant: Thermo Fisher Scientific
A11950.06
A11950.14
A11950.06EA
111
EUR
A11950.06
A11950.14
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine
Purity: 98%
CAS No.: 304-88-1
Molecular Formula: C13H11NO2
Formula Weight: 213.24
Melting Point: 118-120°
Beilstein Reference: 2212449
MDL No.: MFCD00002111
Formel:
C₁₃H₁₁NO₂ Schmelzpunkt: 118…120 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002111 CAS-Nummer: 304-88-1 EINECS: 206-158-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...