1-Dimethylamino-2-nitroethylene 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11637.03
A11637.06
A11637.03EA
38.7
EUR
A11637.03
A11637.06
1-Dimethylamino-2-nitroethylene 98%
1-Dimethylamino-2-nitroethylene
Purity: 98%
CAS No.: 1190-92-7
Molecular Formula: (CH3)2NCH=CHNO2
Formula Weight: 116.12
Storage Temperature: 2°C to 8°C
Melting Point: 101-105°
Beilstein Reference: 1903136
MDL No.: MFCD00051519
Formel:
C₄H₈N₂O₂ Schmelzpunkt: 101…105 °C |
MDL Number:
MFCD00051519 CAS-Nummer: 1190-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...