p-Toluenesulphinic acid sodium salt, (max. 5% H₂O) 97% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Sodium 4-methylbenzenesulphinate, Sodium-p-toluenesulphinate, 4-Methylbenzenesulphinic acid sodium salt, Sodiumtoluene-4-sulphinate
A11430.14
A11430.22
A11430.36
A11430.14EA
37.5
EUR
A11430.14
A11430.22
A11430.36
p-Toluenesulphinic acid sodium salt, (max. 5% H₂O) 97% (Trockengewicht)
p-Toluenesulphinic acid sodium salt
Purity: 97%
CAS No.: 824-79-3
Molecular Formula: C7H7NaO2S
Formula Weight: 178.19
Melting Point:
Formel:
C₇H₇NaO₂S Molekulargewicht: 178.19 g/mol Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149640 CAS-Nummer: 824-79-3 EINECS: 212-538-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...