Cbz-Tyrosinhydrat, (max. 10% H₂O) 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH
A10966.06
A10966.14
A10966.06EA
40.4
EUR
A10966.06
A10966.14
Cbz-Tyrosinhydrat, (max. 10% H₂O) 99%
Cbz-Tyrosinhydrat
Purity: 99%
CAS No.: 1164-16-5
Molecular Formula: C17H17NO5
Formula Weight: 315.33
Storage Temperature: 2°C to 8°C
Melting Point: 57-60°
Beilstein Reference: 2169918
Optical Rotation: +10° (c=3 in dioxane)
MDL No.: MFCD00037180
Formel:
C₁₇H₁₇NO₅ Molekulargewicht: 315.33 g/mol Schmelzpunkt: 57…60 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00037180 CAS-Nummer: 1164-16-5 EINECS: 214-609-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...