Bis-(cyclohexanon)-oxaldihydrazon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone
A10628.14
A10628.22
A10628.14EA
65
EUR
A10628.14
A10628.22
Bis-(cyclohexanon)-oxaldihydrazon ≥98%
Bis(cyclohexanone)oxalyldihydrazone
Purity: 98%
CAS No.: 370-81-0
Molecular Formula: C14H22N4O2
Formula Weight: 278.36
Melting Point: 208-212°
Beilstein Reference: 2388004
MDL No.: MFCD00001659
Formel:
C₁₄H₂₂N₄O₂ Molekulargewicht: 278.36 g/mol Schmelzpunkt: 208…212 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00001659 CAS-Nummer: 370-81-0 EINECS: 206-729-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...