Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
Lieferant: Thermo Fisher Scientific
88481.03
88481.06
88481.03EA
157
EUR
88481.03
88481.06
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
CAS No.: 34830-11-0
Molecular Formula: Eu(C12H14F3O2)3
Formula Weight: 893.68
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 195-198°
MDL No.: MFCD00074803
Formel:
Eu(C₁₂H₁₄F₃O₂)₃ Schmelzpunkt: 195…198 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00074803 CAS-Nummer: 34830-11-0 EINECS: 252-232-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...