Cyclopentadienylvanadium tetracarbonyl ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
81123.02
81123.04
81123.02EA
123
EUR
81123.02
81123.04
Cyclopentadienylvanadium tetracarbonyl ≥97%
Cyclopentadienylvanadium tetracarbonyl
Purity: 97+%
CAS No.: 12108-04-2
Molecular Formula: C9H5O4V
Formula Weight: 228.08
Physical Form: Crystalline
Sensitivity: Air Sensitive
Melting Point: 138°
Boiling Point: 80-100°/0.1mm subl.
UN No.: UN3466
MDL No.: MFCD00014259
Formel:
C₉H₅O₄V Siedepunkt: 80…100 °C (0.1 mmHg) (subl.) Schmelzpunkt: 138 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014259 CAS-Nummer: 12108-04-2 EINECS: 235-163-9 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...