Eisen(III)oxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
47247.A1
47247.18
47247.30
47247.A1EA
860
EUR
47247.A1
47247.18
47247.30
Eisen(III)oxid ≥99,9% (Metall-Basis)
Eisen(III)oxid
Purity: 99.9%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: Powder
Odour: Odourless
Melting Point: 1565°
MDL No.: MFCD00011008
Formel:
Fe₂O₃ Schmelzpunkt: 1565 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011008 CAS-Nummer: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...