Benzyltrimethylammoniumhydroxid 40% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
CAS No.: 100-85-6
Molecular Formula: C10H17NO
Formula Weight: 167.25
Physical Form: Liquid
Density: 1.059
Refractive Index: 1.4300
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Odour: Amine-like
Beilstein Reference: 3917256
UN No.: UN3267
MDL No.: MFCD00008281
Formel:
C₁₀H₁₇NO Dichte: 1.059 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008281 CAS-Nummer: 100-85-6 EINECS: 202-895-5 UN-Nummer: 3267 ADR: 8,II |