Polyoxyethylene bis(amine), M.W. 8,000
Lieferant: Thermo Fisher Scientific
46903.MD
46903.03
46903.06
46903.MDEA
51.7
EUR
46903.MD
46903.03
46903.06
Polyoxyethylene bis(amine), M.W. 8,000
Polyoxyethylene bis(amine)
CAS No.: 24991-53-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2
Physical Form: Granular solid
Sensitivity: Air Sensitive
Appearance: White to off-white
MDL No.: MFCD00132004
Formel:
H₂NCH₂CH₂(OCH₂CH₂)nNH₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00132004 CAS-Nummer: 24991-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...