Polyethylene glycol dimethacrylate, M.W. 3,400
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 25852-47-5
Molecular Formula: CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Storage Temperature: -30°C to -10°C
Physical Form: Granular solid
Sensitivity: Moisture and Light Sensitive
Appearance: White to off-white
MDL No.: MFCD00081877
Formel:
(C₂H₄O)nC₈H₁₀O₃ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00014932 CAS-Nummer: 25852-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...