Aluminiumoxid neutral HPLC Flash-Qualität
Lieferant: Thermo Fisher Scientific
45985.A1
45985.30
45985.A1EA
244
EUR
45985.A1
45985.30
Aluminiumoxid neutral HPLC Flash-Qualität
Aluminiumoxid neutral
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 32-63 Micron APS Powder
Refractive Index: 1.765
Appearance: White
Odour: Odourless
Melting Point: 2045°
Boiling Point: 2980°
MDL No.: MFCD00003424
Formel:
Al₂O₃ Siedepunkt: 2980 °C Schmelzpunkt: 2045 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...