(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dichloro(pentamethylcyclopentadienyl)iridium(III) Dimer, Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, [Ir(Cp-)Cl2]2, (Pentamethylcyclopentadienyl)iridium (III) chloride dimer

45565.04 45565.ME
45565.04EA 877 EUR
45565.04 45565.ME
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

CAS No.: 12354-84-6 Molecular Formula: C20H30Cl4Ir2 Formula Weight: 796.67 MDL No.: MFCD00075435

Formel: C₂₀H₃₀Cl₄Ir₂
Molekulargewicht: 796.69 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00075435
CAS-Nummer: 12354-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR