Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)

Lieferant: Thermo Fisher Scientific

44175.02 44175.77
44175.02EA 310 EUR
44175.02 44175.77
Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)
Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)

CAS No.: 17904-83-5 Molecular Formula: C57H41EuN2O6 Formula Weight: 1001.93 Physical Form: Powder Melting Point: 172-173° Solubility: Soluble in dichloromethane, THF MDL No.: MFCD01321202


Applications: As an organic red light emitter

Formel: C₅₇H₄₁EuN₂O₆
Schmelzpunkt: 172…173 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01321202
CAS-Nummer: 17904-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR