Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtform, geglüht, Ø 0.38 mm (0.015 in) Type 304
Lieferant: Thermo Fisher Scientific
43301.BY
43301.CC
43301.BYEA
50.9
EUR
43301.BY
43301.CC
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtform, geglüht, Ø 0.38 mm (0.015 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Temper: soft
Odour: Odourless
MDL No.: MFCD00148499