Yttrium(III) phosphate hydrate ≥99,99%

Lieferant: Thermo Fisher Scientific
Achtung

41991.04 41991.09
41991.04EA 60.6 EUR
41991.04 41991.09
Yttrium(III) phosphate hydrate ≥99,99%
Yttrium(III) phosphate hydrate

Purity: 99.99% CAS No.: 34054-55-2 Molecular Formula: YPO4.xH2O Formula Weight: 183.88(anhy) Physical Form: Powder MDL No.: MFCD00049782

Formel: YPO₄·xH₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00049782
CAS-Nummer: 34054-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR