Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex -Lösung ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme: Karstedt Catalyst, Platinum(0) - 1,3-Divinyltetramethyldisiloxane Complex
41508.03 41508.06 41508.14
41508.03EA 66.3 EUR
41508.03 41508.06 41508.14
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex -Lösung ≥98%
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex
CAS No.: 68478-92-2 Molecular Formula: (C8H18OSi2)1.5Pt Formula Weight: 474.68 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.984 Sensitivity: Moisture Sensitive Appearance: Orange MDL No.: MFCD00151662
Applications: Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
Formel: C₈H₁₈OPtSi₂
Molekulargewicht: 381.485 g/mol
Dichte: 0.984
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151662
CAS-Nummer: 68478-92-2