Amberlite® IRN-150 Ionenaustauscher
Lieferant: Thermo Fisher Scientific
Synonyme:
Styrene-DVB
40992.36
40992.A3
40992.A9
40992.36EA
72.1
EUR
40992.36
40992.A3
40992.A9
Amberlite® IRN-150 Ionenaustauscher
Amberlite® IRN-150
Molecular Formula: Styrene-DVB
Odour: Odourless
MDL No.: MFCD00145822
Applications: Water treatment and purification
Lagerungstemperatur:
Raumtemperatur |
MDL Number:
MFCD00145822 CAS-Nummer: 79956-14-2 Merck Index: 14,382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...