Strontium molybdenum oxide 99%
Lieferant: Thermo Fisher Scientific
40238.09
40238.18
40238.30
40238.A1
40238.09EA
56.6
EUR
40238.09
40238.18
40238.30
40238.A1
Strontium molybdenum oxide 99%
Strontium molybdenum oxide
Purity: 99.9%
CAS No.: 13470-04-7
Molecular Formula: SrMoO4
Formula Weight: 247.56
Physical Form: -200 Mesh Powder
Density: 4.540
Melting Point: dec.
Solubility: Insoluble in water. Soluble in acid
MDL No.: MFCD00049550
Formel:
SrMoO₄ Schmelzpunkt: Dec. Dichte: 4.540 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00049550 CAS-Nummer: 13470-04-7 EINECS: 236-730-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...