Potassium tris(1-pyrazolyl)borohydride
Lieferant: Thermo Fisher Scientific
39438.03
39438.06
39438.03EA
71.1
EUR
39438.03
39438.06
Potassium tris(1-pyrazolyl)borohydride
Potassium tris(1-pyrazolyl)borohydride
CAS No.: 18583-60-3
Molecular Formula: [C9H10BN6]K
Formula Weight: 252.13
Physical Form: Crystalline
Melting Point: 185-189°
MDL No.: MFCD00040346
Formel:
C₉H₁₀BKN₆ Schmelzpunkt: 185…189 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040346 CAS-Nummer: 18583-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...